Learn More
Electron Microscopy Sciences Cold Embedding Parallel Mold, 30 mm
SDP
Catalog No. 501929689
Encompass_Preferred
Mounting cup with parallel sides that help avoid tilting of the mount when clamped into a specimen holder.
Diameter: 30 mm
Catalog No. 50-192-9689 Supplier Electron Microscopy Sciences Supplier No. 5101275
Mounting cup with parallel sides that help avoid tilting of the mount when clamped into a specimen holder.
Diameter: 30 mm