Learn More
Electron Microscopy Sciences Cold Embedding Parallel Mold, 40 mm
SDP
Catalog No. 501929690
Encompass_Preferred
Mounting cup with parallel sides that help avoid tilting of the mount when clamped into a specimen holder.
Diameter: 40 mm
Catalog No. 50-192-9690 Supplier Electron Microscopy Sciences Supplier No. 5101280
Mounting cup with parallel sides that help avoid tilting of the mount when clamped into a specimen holder.
Diameter: 40 mm