Learn More
Electron Microscopy Sciences Paramat Extra 10 KG
SDP
Catalog No. 5027685
Encompass_Preferred
Paramat is the original British blend of paraffin wax and plastic polymers and it has been used in histology labs throughout the world for over 20 years. It has been found that when plastic polymers are added to the paraffin the elasticity of the final block is greater as compared with paraffin alone. As well the mixture offers improved tissue penetration, easier ribboning of sections, reduced tendency to crumble, and improved overall results with fibrous tissue. Faster penetration with small amount of dimethyl sulfoxide.
Catalog No. 50-276-85 Supplier Electron Microscopy Sciences Supplier No. 1928710
Paramat is the original British blend of paraffin wax and plastic polymers and it has been used in histology labs throughout the world for over 20 years. It has been found that when plastic polymers are added to the paraffin the elasticity of the final block is greater as compared with paraffin alone. As well the mixture offers improved tissue penetration, easier ribboning of sections, reduced tendency to crumble, and improved overall results with fibrous tissue. Faster penetration with small amount of dimethyl sulfoxide.