Learn More
Medchemexpress LLC Paramethasone acetate | 1597-82-6 | 98.0% | 434.50 g/mol | C24H31FO6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10793910MG
Encompass_Preferred
Paramethasone acetate is a synthetic glucocorticoid acetate supplied as a solid research reagent with reported purity of 98% and a molecular weight of 434.50 g/mol. It is used in research to study anti-inflammatory and immunosuppressive effects, glucocorticoid receptor activity, and related signaling pathways. Storage recommendations and an SDS are provided for safe handling and stability considerations.
  • High listed purity suitable for research applications.
  • Supplied as a solid powder for precise dosing and solution preparation.
  • Useful for in vitro and preclinical studies of glucocorticoid activity.
  • Provides storage and stability guidance for powder and solutions.
  • Accompanied by safety data and documentation for laboratory use.
Catalog No. 50-002-42659