Learn More
Medchemexpress LLC Paramethasone acetate | 1597-82-6 | MFCD00200359 | 98.0% | 434.50 g·mol⁻¹ | C24H31FO6 | 5 MG
SDP
Catalog No. 5000241978
Encompass_Preferred
Paramethasone acetate is a synthetic glucocorticoid acetate ester supplied for research use. It shows anti-inflammatory and immunosuppressant activity and is provided as a solid reagent for laboratory and analytical applications. Safety data and handling information are available in the SDS and supporting documentation.
  • Exhibits anti-inflammatory and immunosuppressant activity.
  • Acetate ester form suitable for standard biochemical assays.
  • High purity (98.0%) for reproducible research results.
  • Solid, white to light yellow appearance simplifies storage and handling.
  • Available in small research quantities (e.g., 5 mg).
  • SDS and safety documentation available for laboratory compliance.
Catalog No. 50-002-41978 Supplier Medchemexpress LLC Supplier No. HY1079395MG
Paramethasone acetate is a synthetic glucocorticoid acetate ester supplied for research use. It shows anti-inflammatory and immunosuppressant activity and is provided as a solid reagent for laboratory and analytical applications. Safety data and handling information are available in the SDS and supporting documentation.
  • Exhibits anti-inflammatory and immunosuppressant activity.
  • Acetate ester form suitable for standard biochemical assays.
  • High purity (98.0%) for reproducible research results.
  • Solid, white to light yellow appearance simplifies storage and handling.
  • Available in small research quantities (e.g., 5 mg).
  • SDS and safety documentation available for laboratory compliance.