Learn More
Cayman Chemical paramethoxyButyryl fentnyl
SDP
Catalog No. 502909573
Encompass_Preferred
An internal standard for the quantification of para-methoxy-butyryl fentanyl by GC- or LC-MS; regulated as a Schedule I compound in the United States; intended for research and forensic applications
Catalog No. 50-290-9573 Supplier Cayman Chemical Supplier No. 36588100 UG
An internal standard for the quantification of para-methoxy-butyryl fentanyl by GC- or LC-MS; regulated as a Schedule I compound in the United States; intended for research and forensic applications