Learn More
Meridian Bioscience™ Para-pak SVT
GSA_VA
Catalog No. 23290148 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
120 Vials/Cs.
Catalog No. Quantity
23-290-148 120 Vials/Cs.
1 options
Catalog No. 23-290-148 Supplier Meridian Bioscience™ Supplier No. 901612
Add to Cart
Add to Cart
A convenient one-vial system containing fixative for the recovery of parasites by direct examination, concentration, permanent stain, and in fecal immunoassays.
  • Provides a standardized procedure for the routine collection, transportation, preservation, and examination of stool specimens for intestinal parasites.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Color Green
Quantity 120 Vials/Cs.
Volume (Metric) 15 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov