Learn More
Leica Biosystems Paraplast High Melt High Melt Point 8
Supplier:  Leica Biosystems 39601095
Encompass_Preferred
Paraplast High Melt embedding media is made from a combination of highly purified wax and plastic polymers of regulated molecular weights.Produced by the same manufacturers of Paraplast, Paraplast High Melt can be used for infiltration or embedding and offers reproducibility while sectioning.
Catalog No. 50-255-2390