Learn More
Electron Microscopy Sciences Paraplast Plus 8 KG
SDP
Catalog No. 5027692
Encompass_Preferred
Reduces infiltration time by one-third. Cuts to 2 micron thickness with excellent ribbon continuity. Convenient pellet form melts rapidly. Melting point 56 degree C. Double filtered. No need for filtration before use.
Catalog No. 50-276-92 Supplier Electron Microscopy Sciences Supplier No. 19217
Reduces infiltration time by one-third. Cuts to 2 micron thickness with excellent ribbon continuity. Convenient pellet form melts rapidly. Melting point 56 degree C. Double filtered. No need for filtration before use.