Learn More
Electron Microscopy Sciences Paraplast X-Tra 1 KG
SDP
Catalog No. 5027688
Encompass_Preferred
Melting point: 53-54°C
Handy pellet form melts rapidly. Melting Point 53-54 degree C. Cuts to 2 micron thickness with exceptional ribbon continuity. Extra compression resistance provides total support of tissues
Catalog No. 50-276-88 Supplier Electron Microscopy Sciences Supplier No. 19214
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Melting point: 53-54°C
Handy pellet form melts rapidly. Melting Point 53-54 degree C. Cuts to 2 micron thickness with exceptional ribbon continuity. Extra compression resistance provides total support of tissues