Learn More
Electron Microscopy Sciences Paraplast X-Tra 8 KG
SDP
Catalog No. 5027689
Encompass_Preferred
Melting point: 53-54°C
Handy pellet form melts rapidly. Melting Point 53-54 degree C. Cuts to 2 micron thickness with exceptional ribbon continuity. Extra compression resistance provides total support of tissues
Catalog No. 50-276-89 Supplier Electron Microscopy Sciences Supplier No. 19215
Melting point: 53-54°C
Handy pellet form melts rapidly. Melting Point 53-54 degree C. Cuts to 2 micron thickness with exceptional ribbon continuity. Extra compression resistance provides total support of tissues