Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab ParaPro LMP Paraffin
SDP

Catalog No. 22021367
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-367 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-367 Supplier StatLab Supplier No. PPLMP
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab ParaPro LMP Paraffin - Low Melting Point

  • A medium/hard paraffin
  • For infiltration and embedding
  • Consists of a specialized blend of low-molecular-weight paraffin and polymers to reduce melting point
  • This lower melting point protects tissues from excessive temperature during processing
  • Melting point 50°C to 54°C
  • 1 kg bag, 8/Cs.

Specifications

Physical Form Pellet
Color White
Quantity 1 kg
Grade Histological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.