Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraquat solution, 100μg/mL in water, Ultra Scientific

Catalog No. USPST740AS
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPST740AS 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPST740AS Supplier Agilent Technologies Supplier No. PST740AS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1910-42-5 , 7732-18-5
Molecular Formula C12H14Cl2N2
Molecular Weight (g/mol) 257.16
MDL Number MFCD00150001
InChI Key FIKAKWIAUPDISJ-UHFFFAOYSA-L
PubChem CID 15938
ChEBI CHEBI:28786
IUPAC Name 1,1'-dimethyl-[4,4'-bipyridine]-1,1'-diium dichloride
SMILES [Cl-].[Cl-].C[N+]1=CC=C(C=C1)C1=CC=[N+](C)C=C1

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Water,paraquat (corrected from dichloride)
Solvent or Matrix Water
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Paraquat solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.