Learn More
Cayman Chemical Paraquat (chloride), Purity: ≥95%, CAS: 1910-42-5, 100 mg
SDP
Catalog No. 502917131
Encompass_Preferred
An herbicide; induces light-dependent apoptosis and reduces chlorophyll levels in tobacco leaf discs at 10 µM; eliminates susceptible barley grass but not the paraquat-resistant biotype in field studies at 0.05, 0.1, and 0.2 kg/ha; induces apoptosis, increases intracellular levels of NO, and increases expression of mRNA encoding iNOS in SH-SY5Y cells at 100 µM; toxic to rats (LD50s = 115 and 21 mg/kg for p.o. and i.v. administration, respectively); induces neuronal cell death and seizures, effects that can be inhibited by U-74389G, in rats at 100 nmol/animal, i.c.v.; induces weight loss, lung hemorrhage and edema, and diarrhea in rats at 20 mg/kg
Catalog No. 50-291-7131 Supplier Cayman Chemical Supplier No. 38521100 MG
An herbicide; induces light-dependent apoptosis and reduces chlorophyll levels in tobacco leaf discs at 10 µM; eliminates susceptible barley grass but not the paraquat-resistant biotype in field studies at 0.05, 0.1, and 0.2 kg/ha; induces apoptosis, increases intracellular levels of NO, and increases expression of mRNA encoding iNOS in SH-SY5Y cells at 100 µM; toxic to rats (LD50s = 115 and 21 mg/kg for p.o. and i.v. administration, respectively); induces neuronal cell death and seizures, effects that can be inhibited by U-74389G, in rats at 100 nmol/animal, i.c.v.; induces weight loss, lung hemorrhage and edema, and diarrhea in rats at 20 mg/kg