Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pararosaniline chloride, 96%, pure

Catalog No. AC416800250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416800250 25 g Glass bottle
AC416800050 5 g Glass bottle
AC416801000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC416800250 Supplier Thermo Scientific Chemicals Supplier No. 416800250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 569-61-9 | C19H17N3·HCl | 323.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H17N3·HCl
Molecular Weight (g/mol) 323.83
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 268.0°C to 270.0°C
Color Green
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol
Formula Weight 323.83
Percent Purity 98%
Grade Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Pararosaniline chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-321-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.