Learn More
Medchemexpress LLC Pararosaniline hydrochloride | 569-61-9 | 00001657 | 97.3% | 323.83 g/mol | C19H18ClN3 | 10 G
SDP
Catalog No. 5000282741
Encompass_Preferred
Pararosaniline hydrochloride is a basic red dye provided as a solid powder for laboratory staining and analytical applications. It is used as a biological stain and in Schiff reagent formulations, exhibits pH-responsive color behavior, and is supplied at high purity in small pack sizes suitable for bench-top use.
  • Basic red dye for histological and cytological staining.
  • pH-responsive color change useful in analytical assays.
  • Solid powder form, easy to weigh and dissolve.
  • High purity suitable for laboratory applications.
  • Available in small pack sizes for routine use.
Catalog No. 50-002-82741 Supplier Medchemexpress LLC Supplier No. HYW12777010G
Pararosaniline hydrochloride is a basic red dye provided as a solid powder for laboratory staining and analytical applications. It is used as a biological stain and in Schiff reagent formulations, exhibits pH-responsive color behavior, and is supplied at high purity in small pack sizes suitable for bench-top use.
  • Basic red dye for histological and cytological staining.
  • pH-responsive color change useful in analytical assays.
  • Solid powder form, easy to weigh and dissolve.
  • High purity suitable for laboratory applications.
  • Available in small pack sizes for routine use.