Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pararosaniline Hydrochloride, Sigma-Aldrich

Catalog No. 01258820 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-820 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-820 Supplier MilliporeSigma Supelco Supplier No. P375025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pararosaniline hydrochloride has been used:

  • to prepare Schiff reagent
  • to stain cow endometrial cytology samples
  • to counterstain Prussian blue sections of mice brains
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-61-9
Molecular Weight (g/mol) 323.82 g/mol

Specifications

Color Black to Green
Quantity 25 g
Percent Purity >85%
Chemical Name or Material Pararosaniline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.