Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pararot, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015036 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5036 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5036 Supplier MilliporeSigma Supelco Supplier No. 40446100MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 6410-10-2
Physical Form Neat
Linear Formula O2 NC6H4 N=NC10H6 OH
MDL Number MFCD00003908
Melting Point 248°C to 252°C
Synonym Para Red; 1-(4-Nitrophenylazo)-2-naphthol; 1-(p-Nitrophenylazo)-2-naphthol
Type Analytical Standard
Quantity 100 mg
Formula Weight 293.28
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.