Learn More
Medchemexpress LLC HY-P0324A 1mg Medchemexpress, Parasin I (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0324A1MG
Encompass_Preferred
Medchemexpress, HY-P0324A 1mg Parasin I (TFA) CAS: Parasin I (TFA) is a 19-amino acid histone H2A-derived peptide isolated from the skin of the catfish, and shows antimicrobial activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4119