Learn More
Medchemexpress LLC Parasin I TFA | 219552-69-9 | 99.8% | 2114.33 | 5 MG
SDP
Catalog No. 5000422146
Encompass_Preferred
Parasin I (TFA) is a 19-amino acid histone H2A-derived peptide isolated from the skin of the catfish, exhibiting antimicrobial activity. It is intended for research use only and not for patient administration.
  • 19-amino acid histone H2A-derived peptide
  • Isolated from the skin of catfish
  • Shows antimicrobial activity
  • White to off-white solid appearance
  • Sequence: Lys-Gly-Arg-Gly-Lys-Gln-Gly-Gly-Lys-Val-Arg-Ala-Lys-Ala-Lys-Thr-Arg-Ser-Ser
  • Initial source: Animals, the skin mucus of wounded catfish
Catalog No. 50-004-22146 Supplier Medchemexpress LLC Supplier No. HYP0324A5MG
Parasin I (TFA) is a 19-amino acid histone H2A-derived peptide isolated from the skin of the catfish, exhibiting antimicrobial activity. It is intended for research use only and not for patient administration.
  • 19-amino acid histone H2A-derived peptide
  • Isolated from the skin of catfish
  • Shows antimicrobial activity
  • White to off-white solid appearance
  • Sequence: Lys-Gly-Arg-Gly-Lys-Gln-Gly-Gly-Lys-Val-Arg-Ala-Lys-Ala-Lys-Thr-Arg-Ser-Ser
  • Initial source: Animals, the skin mucus of wounded catfish