Learn More
Cayman Chemical ParasIn IcatfIshtrIfluor 10mg
SDP
Catalog No. 502910042
Encompass_Preferred
An antimicrobial peptide; active against various Gram-positive and Gram-negative bacteria, as well as fungi (MICs = 1-2, 1-4, and 1-2 UG/ml, respectively)
Catalog No. 50-291-0042 Supplier Cayman Chemical Supplier No. 3672410 MG
An antimicrobial peptide; active against various Gram-positive and Gram-negative bacteria, as well as fungi (MICs = 1-2, 1-4, and 1-2 UG/ml, respectively)