Learn More
Electron Microscopy Sciences Parasite Magnifier
SDP
Supplier:  Electron Microscopy Sciences 68110
Encompass_Preferred
An improved version of the conventional magnifying glass. A 20 mm diameter parasite-like lens is cemented on a large 75 mm diameter lens. Magnification of the large one is 2.5X, while the small lens area has the magnification of 5X. The magnified image is very sharp and clear. Weight: 111g
Catalog No. 50-276-94