Learn More
Alpco Ltd Parasitic Stool Panel I PCR Kit
SDP
Catalog No. NC2284610
Encompass
The Parasitic Stool Panel I is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of Giardia lamblia, Cryptosporidium spp., Entamoeba histolytica and Dientamoeba fragilis in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284610 Supplier Alpco Ltd Supplier No. 86PS1HU100
The Parasitic Stool Panel I is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of Giardia lamblia, Cryptosporidium spp., Entamoeba histolytica and Dientamoeba fragilis in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.