Learn More
Alpco Ltd Parasitic Stool Panel II PCR Kit
SDP
Catalog No. NC2284611
Encompass
The Parasitic Stool Panel II is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of Giardia lamblia, Cryptosporidium spp. and Entamoeba histolytica in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.
This item is not returnable. View return policy
Catalog No. NC2284611 Supplier Alpco Ltd Supplier No. 86PS2HU100
The Parasitic Stool Panel II is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of Giardia lamblia, Cryptosporidium spp. and Entamoeba histolytica in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures. This kit is sufficient for 100 tests.