Learn More
Volu Sol Parasitology Kit (Wheatley's Procedure), Refill, each, Volu-Sol™
SDP
Catalog No. 50420827 Shop All Volu Sol Products
Encompass_Preferred
Single package containing all the necessary materials and reagents to prepare stained fecal smears.  Provides excellent stain results differentiating internal structures of intestinal parasites as well as facilitating the separation of these organisms from background material and artifacts
Catalog No. 50420827 Supplier Volu Sol Supplier No. VXR010
Single package containing all the necessary materials and reagents to prepare stained fecal smears.  Provides excellent stain results differentiating internal structures of intestinal parasites as well as facilitating the separation of these organisms from background material and artifacts