Learn More
Chem Service Inc Parathion, 56-38-2, MFCD00036219, 100 mg
SDP
Supplier:  Chem Service Inc N12819100MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula: C10-H14-N-O5-P-S, Molecular Weight: 291.26, Classification: Insecticide, Acaricide, Alternate Name(s): O.O-Diethyl-O-p-nitrophenyl phosphorothioate.
Catalog No. NC1204281