Learn More
Cayman Chemical ParathyroId Hormon1-13t 500ug
SDP
Catalog No. 502933012
Encompass_Preferred
An N-terminal fragment of PTH; increases MAP in anesthetized rats from 0.003-1 μmol/kg
Catalog No. 50-293-3012 Supplier Cayman Chemical Supplier No. 24984500 UG
An N-terminal fragment of PTH; increases MAP in anesthetized rats from 0.003-1 μmol/kg