Learn More
Cayman Chemical ParathyroId Hormon1-34r 1mg
SDP
Catalog No. 502909757
Encompass_Preferred
A synthetic N-terminal fragment of PTH and an agonist of PTH1R and PTH2R; induces β-arrestin recruitment in HEK293H cells expressing human PTH receptor type 1 (PTH1R; EC50 = 7.7 nM) and adenylyl cyclase activation in COS-7 cells expressing rat PTH2R (EC50 = 0.41 nM); induces hypertrophy of primary rat ventricular cardiomyocytes at 100 nM; increases tibial bone mineral density in ovariectomized rats at 5 UG/kg
Catalog No. 50-290-9757 Supplier Cayman Chemical Supplier No. 368611 MG
A synthetic N-terminal fragment of PTH and an agonist of PTH1R and PTH2R; induces β-arrestin recruitment in HEK293H cells expressing human PTH receptor type 1 (PTH1R; EC50 = 7.7 nM) and adenylyl cyclase activation in COS-7 cells expressing rat PTH2R (EC50 = 0.41 nM); induces hypertrophy of primary rat ventricular cardiomyocytes at 100 nM; increases tibial bone mineral density in ovariectomized rats at 5 UG/kg