Learn More
Sigma Aldrich Fine Chemicals Biosciences Parathyroid Hormone 1 84 H
Catalog No. 502905304
Encompass
Parathyroid hormone (PTH) or parathormone is secreted by the parathyroid glands as a polypeptide containing 84 amino acids. It acts to increase the concentration of calcium in the blood whereas calcitonin (a hormone produced by the parafollicular cells of the thyroid gland) acts to decrease calcium concentration. Parathyroid Hormone 1-84 (full length) Human Recombinant produced in E. coli is a single non-glycosylated polypeptide chain containing 84 amino acids (MW of 9.4kDa) with the following amino acid sequenceSVSEIQLMHN LGKHLNSMER VEWLRKKLQD VHNfVALGAP LAPRDAGSQR PRKKEDNVLV ESHEKSLGEA DKADVNVLTK AKSQ
Catalog No. 50-290-5304 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE0192100UG
Parathyroid hormone (PTH) or parathormone is secreted by the parathyroid glands as a polypeptide containing 84 amino acids. It acts to increase the concentration of calcium in the blood whereas calcitonin (a hormone produced by the parafollicular cells of the thyroid gland) acts to decrease calcium concentration. Parathyroid Hormone 1-84 (full length) Human Recombinant produced in E. coli is a single non-glycosylated polypeptide chain containing 84 amino acids (MW of 9.4kDa) with the following amino acid sequenceSVSEIQLMHN LGKHLNSMER VEWLRKKLQD VHNfVALGAP LAPRDAGSQR PRKKEDNVLV ESHEKSLGEA DKADVNVLTK AKSQ