Learn More
Apexbio Technology LLC parathyroid hormone (7-34) [Homo sapiens]/[Macaca fascicularis] 25mg
SDP
Catalog No. 5000096693
Encompass_Preferred
Parathyroid hormone (7-34) is a polypeptide antagonist targeting parathyroid hormone receptors (PTH1R and PTH2R) It is derived from Homo sapiens and Macaca fascicularis and is designed to modulate PTH receptor activity thereby influencing calcium and phosphate homeostasis Parathyroid hormone (7-34) exerts its biological activity primarily through competitive interaction with PTH receptors affecting absorption processes in bone kidney and intestinal tissues Based on these pharmacological properties parathyroid hormone (7-34) holds research potential in studies of mineral metabolism bone remodeling and calcium-phosphate balance disorders
Catalog No. 50-000-96693 Supplier Apexbio Technology LLC Supplier No. A108425
Parathyroid hormone (7-34) is a polypeptide antagonist targeting parathyroid hormone receptors (PTH1R and PTH2R) It is derived from Homo sapiens and Macaca fascicularis and is designed to modulate PTH receptor activity thereby influencing calcium and phosphate homeostasis Parathyroid hormone (7-34) exerts its biological activity primarily through competitive interaction with PTH receptors affecting absorption processes in bone kidney and intestinal tissues Based on these pharmacological properties parathyroid hormone (7-34) holds research potential in studies of mineral metabolism bone remodeling and calcium-phosphate balance disorders