Learn More
CPC Scientific 5FAM-Ser-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PTHP015A
Encompass
This is a fluorescent (FAM)-labeled PTH peptide, Abs/Em=492/518 nm.ONE-LETTER SEQUENCE: 5FAM-SVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNFMOLECULAR FORMULA: C202H301N55O57S2MOLECULAR WEIGHT:4476.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Osteoporosis
Catalog No. 50-211-0354