Learn More
CPC Scientific H-Ala-Val-Ser-Glu-Ile-Gln-Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Ala-Ser-Val-Glu-Arg-Met-Gln-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PTHP011B
Encompass
ONE-LETTER SEQUENCE: AVSEIQLMHNLGKHLASVERMQWLRKKLQDVHNFMOLECULAR FORMULA: C180H291N55O48S2MOLECULAR WEIGHT:4057.76STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [98614-76-7], SYNONYMS: pTH (1-34) (rat)RESEARCH AREA: OsteoporosisREFERENCES: H.T. Keutmann et al., Abstract A. S. B. M. R. Conf. (1984); H.T. Keutmann et al., Endocrinol., 117, 1230 (1985)
Catalog No. 50-211-0350