Learn More
StatLab Parawash Fecal Concentrator Kit
SDP
Catalog No. 22130482
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
22-130-482 Non-sterile
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-482 Supplier StatLab Supplier No. 641
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Concentrate intestinal parasites from preserved fecal specimens with the Fecal Concentrator Kit while reducing processing time.
  • Uniform filter diameter of 0.6 mm allows eggs, larvae, cysts, oocysts, and spores while stopping macroscopic debris
  • Kit contains a 250 mL bottle Parawash, a 15 mL bottle mucolytic digesting agent; 50 - 15 mL polypropylene centrifuge tubes, 50 standardized fine mesh filter funnels, and 50 tube caps
  • 1 kit
Specifications
Product Type Collection and Transport System
Format Tube
Sterility Non-sterile