Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Paraxanthine-d6 | 117490-41-2 | >99.0% | 186.20 | C7H2D6N4O2 | 1 MG
SDP

Catalog No. 5000301141
Encompass_Preferred

Paraxanthine-d6 is the deuterium-labeled analogue of paraxanthine supplied as a stable isotope reference material for analytical and research use. It is intended as an internal standard for quantitative mass spectrometry and tracer studies of caffeine metabolites. The material is provided as a crystalline solid with high isotopic purity and is compatible with common organic solvents.

  • Stable isotope internal standard for quantitative mass spectrometry.
  • High isotopic purity (>99% deuterated forms) for accurate quantitation.
  • Compatible with LC-MS and tracer studies.
  • Crystalline solid, soluble in DMSO and common organic solvents.
  • Molecular formula C7H2D6N4O2; molecular weight 186.20.

Catalog No. 50-003-01141 Supplier Medchemexpress LLC Supplier No. HYW016498S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Paraxanthine-d6 is the deuterium-labeled analogue of paraxanthine supplied as a stable isotope reference material for analytical and research use. It is intended as an internal standard for quantitative mass spectrometry and tracer studies of caffeine metabolites. The material is provided as a crystalline solid with high isotopic purity and is compatible with common organic solvents.

  • Stable isotope internal standard for quantitative mass spectrometry.
  • High isotopic purity (>99% deuterated forms) for accurate quantitation.
  • Compatible with LC-MS and tracer studies.
  • Crystalline solid, soluble in DMSO and common organic solvents.
  • Molecular formula C7H2D6N4O2; molecular weight 186.20.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.