Learn More
Cayman Chemical Parbendazole, Purity: ≥98%, CAS: 14255-87-9, 100 mg
SDP
Catalog No. 502910771
Encompass_Preferred
An inhibitor of microtubule assembly (EC50 = 3 µM); reduces viability of HeLa cells (EC50 = 0.53 µM); induces genotoxic stress in a reporter assay using HEK293 cells; reduces, but does not eradicate, worm burden in goats with naturally acquired or experimental nematode infections at 15 mg/kg; teratogenic to lamb embryos when administered to pregnant ewes at 60 mg/kg during the third week of pregnancy
Catalog No. 50-291-0771 Supplier Cayman Chemical Supplier No. 34754100 MG
An inhibitor of microtubule assembly (EC50 = 3 µM); reduces viability of HeLa cells (EC50 = 0.53 µM); induces genotoxic stress in a reporter assay using HEK293 cells; reduces, but does not eradicate, worm burden in goats with naturally acquired or experimental nematode infections at 15 mg/kg; teratogenic to lamb embryos when administered to pregnant ewes at 60 mg/kg during the third week of pregnancy