Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterprise Technology Solutions Parchment Specialty Paper 24 l
SDP

Catalog No. 502884852
Encompass_Preferred

Parchment Specialty Paper 24 lb Bond Weight 8.5 x 11 Blue 500/Ream

Catalog No. 50-288-4852 Supplier Enterprise Technology Solutions Supplier No. SOU964C
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Parchment Specialty Paper 24 lb Bond Weight 8.5 x 11 Blue 500/Ream

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.