Learn More
Aviva Systems Biology Corp PARD3 Antibody - middle region (ARP42910_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42910_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PARD3. Alternate names: ASIP, Baz, Bazooka, FLJ21015, PAR3, PAR3alpha, PARD3A, SE2-5L16, SE2-5LT1, SE2-5T2ApplicationWB, IHC. Species reactivity: Guinea Pig, Human
Catalog No. 50-014-58358