Learn More
Novus Biologicals™ PARD3/Par3 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 56288 |
| Species | Mouse |
| Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP2-11253 |
| Gene Alias | FLJ21015, par-3 partitioning defective 3 homolog (C. elegans), PAR3A, PAR3C.elegans) homolog |
| Gene Symbol | PARD3 |
| Quantity | 0.25 mg |
| Research Category | Cell Cycle and Replication |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.