Learn More
Aviva Systems Biology Corp PARD6A Antibody - N-terminal region (ARP42883_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42883_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARD6A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PARD6A. Alternate names: PAR-6A, PAR6, PAR6C, PAR6alpha, TAX40, TIP-40ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53890