Learn More
Medchemexpress LLC HY-14958A 5mg Medchemexpress, Pardoprunox (hydrochloride) CAS:269718-83-4 Purity:>98%
SDP
Catalog No. 502081668
Encompass_Preferred
Medchemexpress, HY-14958A 5mg Pardoprunox (hydrochloride) CAS:269718-83-4 Pardoprunox (SLV-308) hydrochloride is a partial dopamine D2 and D3 receptor partial agonist and a serotonin 5-HT1A receptor agonist, with pEC50s of 8, 9.2, and 6.3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1668 Supplier Medchemexpress LLC Supplier No. HY14958A5MG
Medchemexpress, HY-14958A 5mg Pardoprunox (hydrochloride) CAS:269718-83-4 Pardoprunox (SLV-308) hydrochloride is a partial dopamine D2 and D3 receptor partial agonist and a serotonin 5-HT1A receptor agonist, with pEC50s of 8, 9.2, and 6.3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.