Learn More
Medchemexpress LLC Pardoprunox hydrochloride | 269718-83-4 | 99.5% | 269.73 | 100 MG
SDP
Catalog No. 5000360015
Encompass_Preferred
Pardoprunox hydrochloride is a compound that acts as a partial agonist for both dopamine D2 and D3 receptors, and a full agonist for the serotonin 5-HT1A receptor. This compound is intended for research purposes only.
  • Partial dopamine D2 and D3 receptor agonist
  • Serotonin 5-HT1A receptor agonist
  • Acts as a partial D2 receptor agonist with an efficacy of 50%
  • Induces [(35)S]GTPgammaS binding as a partial D3 receptor agonist
  • Full 5-HT1A receptor agonist on forskolin induced cAMP accumulation
Catalog No. 50-003-60015 Supplier Medchemexpress LLC Supplier No. HY14958A100MG
Pardoprunox hydrochloride is a compound that acts as a partial agonist for both dopamine D2 and D3 receptors, and a full agonist for the serotonin 5-HT1A receptor. This compound is intended for research purposes only.
  • Partial dopamine D2 and D3 receptor agonist
  • Serotonin 5-HT1A receptor agonist
  • Acts as a partial D2 receptor agonist with an efficacy of 50%
  • Induces [(35)S]GTPgammaS binding as a partial D3 receptor agonist
  • Full 5-HT1A receptor agonist on forskolin induced cAMP accumulation