Learn More
Medchemexpress LLC Pardoprunox hydrochloride | 269718-83-4 | 99.5% | 269.73 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14958A
Encompass_Preferred
Pardoprunox hydrochloride (SLV-308) is a partial dopamine D2 and D3 receptor partial agonist and a serotonin 5-HT1A receptor agonist. It demonstrates potent partial agonism at D2 receptors and partial agonism at D3 receptors, while also acting as a full agonist for 5-HT1A receptors.
  • Partial dopamine D2 and D3 receptor agonist
  • Serotonin 5-HT1A receptor agonist
  • Potent partial D2 receptor agonist with 50% efficacy on forskolin stimulated cAMP accumulation
  • Partial agonist at human recombinant dopamine D3 receptors, inducing [(35)S]GTPgammaS binding
  • Full 5-HT1A receptor agonist on forskolin induced cAMP accumulation
Catalog No. 50-003-63961