Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pardoprunox hydrochloride | 269718-83-4 | MFCD09833663 | 99.5% | 269.73 g/mol | C12H16ClN3O2 | 10 MG
SDP

Catalog No. 5000286067
Encompass_Preferred

Pardoprunox hydrochloride is the hydrochloride salt of SLV-308, a research compound used in pharmacology studies of dopaminergic and serotonergic signaling. It is provided as a high-purity solid and formulated for both in vitro and in vivo experiments.

  • Acts as a partial agonist at dopamine D2 and D3 receptors and an agonist at 5-HT1A receptors.
  • Reported pEC50 values: D2 = 8, D3 = 9.2, 5-HT1A = 6.3.
  • Supplied as an off-white to pink solid suitable for laboratory use.
  • High purity appropriate for research applications.
  • Soluble in DMSO and water; in vivo formulations and concentrations provided for dosing.
  • Storage recommendations: solid at 4°C; dissolved samples at -80°C for long-term storage.

Catalog No. 50-002-86067 Supplier Medchemexpress LLC Supplier No. HY14958A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pardoprunox hydrochloride is the hydrochloride salt of SLV-308, a research compound used in pharmacology studies of dopaminergic and serotonergic signaling. It is provided as a high-purity solid and formulated for both in vitro and in vivo experiments.

  • Acts as a partial agonist at dopamine D2 and D3 receptors and an agonist at 5-HT1A receptors.
  • Reported pEC50 values: D2 = 8, D3 = 9.2, 5-HT1A = 6.3.
  • Supplied as an off-white to pink solid suitable for laboratory use.
  • High purity appropriate for research applications.
  • Soluble in DMSO and water; in vivo formulations and concentrations provided for dosing.
  • Storage recommendations: solid at 4°C; dissolved samples at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.