Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Parecoxib (Sodium) | 198470-85-8 | 99.6% | 392.40 | 10 MM 1 ML
SDP

Catalog No. 5000386879
Encompass_Preferred

Parecoxib Sodium is a highly selective, orally active, and blood-brain barrier-permeable COX-2 inhibitor, functioning as a prodrug of Valdecoxib. It is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits prostaglandin synthesis. It is used for the relief of acute postoperative pain and symptoms of chronic inflammatory conditions like osteoarthritis and rheumatoid arthritis.

  • Target: COX-2
  • Inhibits cell proliferation of GBM cells in a dose-dependent manner
  • Decreases migratory ability of U343 cells
  • Chronic administration exerts anxiolytic-like and memory-enhancing effects in mice
  • Modulates synaptophysin expression in mice
  • Appearance: Solid
  • Color: White to off-white
  • Shipping at room temperature in continental US
  • Storage at 4°C, sealed, away from moisture
  • In solvent storage at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-86879 Supplier Medchemexpress LLC Supplier No. HY17474A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Parecoxib Sodium is a highly selective, orally active, and blood-brain barrier-permeable COX-2 inhibitor, functioning as a prodrug of Valdecoxib. It is a nonsteroidal anti-inflammatory agent (NSAID) that inhibits prostaglandin synthesis. It is used for the relief of acute postoperative pain and symptoms of chronic inflammatory conditions like osteoarthritis and rheumatoid arthritis.

  • Target: COX-2
  • Inhibits cell proliferation of GBM cells in a dose-dependent manner
  • Decreases migratory ability of U343 cells
  • Chronic administration exerts anxiolytic-like and memory-enhancing effects in mice
  • Modulates synaptophysin expression in mice
  • Appearance: Solid
  • Color: White to off-white
  • Shipping at room temperature in continental US
  • Storage at 4°C, sealed, away from moisture
  • In solvent storage at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.