Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Parecoxib sodium impurity 10 | 198471-06-6 | 97.0% | 356.40 | C18H16N2O4S | 250 MG
SDP

Catalog No. 5000303538
Encompass_Preferred

Parecoxib sodium impurity 10 is a characterized impurity of parecoxib sodium supplied as a research reference standard for analytical, synthetic, and impurity-identification work. It is provided for research use only and not for human or clinical applications.

  • Used as an analytical reference for impurity profiling and method development.
  • Intended for use as a drug intermediate or impurity characterization standard.
  • Purity: 97.0% (as listed by supplier).
  • Molecular formula C18H16N2O4S; molecular weight 356.40.
  • Available in multiple sizes, including 250 mg, to support R&D and analytical needs.
  • Store under conditions specified in the certificate of analysis to ensure stability.

Catalog No. 50-003-03538 Supplier Medchemexpress LLC Supplier No. 250MGHYW038820
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Parecoxib sodium impurity 10 is a characterized impurity of parecoxib sodium supplied as a research reference standard for analytical, synthetic, and impurity-identification work. It is provided for research use only and not for human or clinical applications.

  • Used as an analytical reference for impurity profiling and method development.
  • Intended for use as a drug intermediate or impurity characterization standard.
  • Purity: 97.0% (as listed by supplier).
  • Molecular formula C18H16N2O4S; molecular weight 356.40.
  • Available in multiple sizes, including 250 mg, to support R&D and analytical needs.
  • Store under conditions specified in the certificate of analysis to ensure stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.