Learn More
Cayman Chemical Parecoxib (sodium salt), Purity: ≥98%, CAS: 198470-85-8, 100 mg
SDP
Catalog No. 502938207
Encompass_Preferred
A prodrug form of valdecoxib; converted to valdecoxib by human liver microsomes in vitro and in vivo in rats, dogs, and cynomolgous monkeys; a CB1 agonist (EC50 = 2.4 µM in HEK293 cells); reduces hyperalgesia in a rat model of carrageenan-induced foot pad edema (ED50 = 5 mg/kg) and decreases inflammation in a rat model of M. butyricum-induced arthritis (ED50 = 0.08 mg/kg)
Catalog No. 50-293-8207 Supplier Cayman Chemical Supplier No. 29017100 MG
A prodrug form of valdecoxib; converted to valdecoxib by human liver microsomes in vitro and in vivo in rats, dogs, and cynomolgous monkeys; a CB1 agonist (EC50 = 2.4 µM in HEK293 cells); reduces hyperalgesia in a rat model of carrageenan-induced foot pad edema (ED50 = 5 mg/kg) and decreases inflammation in a rat model of M. butyricum-induced arthritis (ED50 = 0.08 mg/kg)