Learn More
Med Vet International Calplus Parenteral Solution, 500mL, Each
Catalog No. 502828877
Encompass_Preferred
Parenteral solution for intravenous use in cattle to treat milk fever, and to treat calcium, phosphorus, magnesium and glucose deficiency in cattle, horses and sheep. Contains calcium borogluconate, dextrose, magnesium borogluconate, calcium hypophosphite and base.
Catalog No. 50-282-8877 Supplier Med Vet International Supplier No. RXCAL500INJV
Parenteral solution for intravenous use in cattle to treat milk fever, and to treat calcium, phosphorus, magnesium and glucose deficiency in cattle, horses and sheep. Contains calcium borogluconate, dextrose, magnesium borogluconate, calcium hypophosphite and base.