Learn More
Electron Microscopy Sciences Paraformaldehyde 1% with Glutaraldehyde 2% in 0.1M Phosphate Buffer, pH 7.4 500 ML
SDP
Supplier:  Electron Microscopy Sciences 1594935
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. Paraformaldehyde 1 % Glutaraldehyde 2% in 0.1M Phosphate Buffer.
pH: 7.4
Catalog No. 50-190-1152