Learn More
Electron Microscopy Sciences Paraformaldehyde 4% in 0.1M Phosphate Buffer, pH 7.4 10 ML
SDP
Catalog No. 501901158
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Phosphate Buffer. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4
Catalog No. 50-190-1158 Supplier Electron Microscopy Sciences Supplier No. 1573510S
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Phosphate Buffer. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4