Learn More
Electron Microscopy Sciences Paraformaldehyde 4 % in 0.1M Sodium Cacodylate Buffer, pH 7.4 10 ML
SDP
Catalog No. 501901156
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Sodium Cacodylate Buffer. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4
Catalog No. 50-190-1156 Supplier Electron Microscopy Sciences Supplier No. 1595210 S
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.1M Sodium Cacodylate Buffer. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4